Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ Corrugated Waste Gas Tubing
SDP

Catalog No. 04777188 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
04-777-188 Research Animal Inhalant Anesthesia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-777-188 Supplier SOMNI Scientific™ Supplier No. TB2008

missing translation for 'validMainframeMsg'

Provided custom cut per 12-inch increments to your specification. This flexible scavenger tubing is color coded purple for easy identification.

  • 19 mm Corrugated Waste Gas Tubing is provided custom cut per 12 inch increments to your specification.
  • Flexible scavenger tubing
  • Color coded purple for easy identification
  • Applications include: WAG filters, active WAG systems, rebreathing systems, induction chambers, nosecones

Specifications

For Use With (Application) Research Animal Inhalant Anesthesia
Length (Metric) 19 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.