Learn More
Aviva Systems Biology Corp CORT Antibody - N-terminal region (ARP59541_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59541_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CORT. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CORT. Alternate names: CST-14, CST-17, CST-29ApplicationWB. Species reactivity: Human
Catalog No. 50-286-734