Learn More
Waters Corp Waters CORTECS C18 Column, 90Å, 2.7 μm, 2.1 × 100 mm
Supplier:  Waters Corp 186007367
Encompass_Preferred
CORTECS C18 Columns are general purpose, high-efficiency columns built on a solid-core particle. They provide balanced retention for acids, bases, and neutrals across low to mid-range pH levels, ensuring maximum efficiency and enhanced resolution for complex samples. This technology allows for increased sample throughput and decreased analytical costs.
  • General purpose, high-efficiency columns with solid-core particle technology
  • Offers balanced retention for acids, bases, and neutrals at low to mid-range pH
  • Provides maximum efficiency and resolution for complex sample analysis
  • Increases sample throughput with higher flow rates
  • Reduces analytical and operational costs
  • Allows shorter column lengths with maintained peak capacities and faster re-equilibration
  • Ensures consistent performance and reproducibility through tightly monitored manufacturing
Catalog No. 50-592-601