Learn More
Waters Corp Waters Column, C18, 1.6 μm, 2.1 mm × 150 mm, 90 Å
Supplier:  Waters Corp 186007096
Encompass_Preferred
These general purpose, high-efficiency columns are based on a solid-core particle that offer balanced retention of acids, bases, and neutrals at low and mid-range pH. The solid-core particle technology maximizes efficiency for maximum resolution, enabling increased sample throughput and faster re-equilibration times.
  • Maximizes efficiency for maximum resolution.
  • Provides increased resolution for analyzing complex samples compared to fully porous columns.
  • Enables increased sample throughput with comparable or better peak capacities.
  • Allows use of shorter column lengths while maintaining similar peak capacities.
  • Offers faster re-equilibration time.
  • Decreases cost of analysis and lab operations.
  • Ensures unparalleled reproducibility and consistent performance through tightly monitored manufacturing.
Catalog No. 50-597-066