Learn More
Waters Corp Waters CORTECS C18+ Column, 90Å, 1.6 μm, 2.1 mm x 150 mm
Supplier:  Waters Corp 186007117
Encompass_Preferred
CORTECS C18+ columns are high-efficiency C18 columns with solid-core particles, designed for excellent peak shape for basic analytes at low pH. These reversed-phase columns improve performance by lowering the van Deemter equation's three terms, leading to increased sample throughput and reduced lab operating costs.
  • High-efficiency C18 columns with solid-core particle design.
  • Delivers excellent peak shape for basic analytes at low pH.
  • Solid core particles improve performance by lowering van Deemter equation terms.
  • Increased sample throughput via higher method flow rates.
  • Shorter column lengths maintain peak capacities, reducing operating costs.
Catalog No. 50-597-086