Learn More
Waters Corp Waters Column, C18, 1.6 μm, 2.1 mm, 30 mm
Catalog No. 50597062 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These general purpose, high-efficiency columns are based on a solid-core particle, offering balanced retention of acids, bases, and neutrals at low and mid-range pH.
  • General purpose, high-efficiency columns based on a solid-core particle.
  • Offer balanced retention of acids, bases and neutrals at low and mid-range pH.
Catalog No. 50-597-062 Supplier Waters Corp Supplier No. 186007092
These general purpose, high-efficiency columns are based on a solid-core particle, offering balanced retention of acids, bases, and neutrals at low and mid-range pH.
  • General purpose, high-efficiency columns based on a solid-core particle.
  • Offer balanced retention of acids, bases and neutrals at low and mid-range pH.