Learn More
Waters Corp Waters Corporation CORTECS UPLC Columns, C18+, 1.6 μm, 2.1 × 30 mm
Catalog No. 50597082 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This CORTECS UPLC Column is a solid-core particle column designed to provide the highest column efficiencies when used with low-dispersive UPLC instrumentation.
  • Offers flexibility to rapidly separate a wide array of compounds.
  • Produces sharper, narrower peaks compared to fully porous particles of similar size.
  • Best column choice for increased resolution, speed, and sensitivity.
Catalog No. 50-597-082 Supplier Waters Corp Supplier No. 186007113
This CORTECS UPLC Column is a solid-core particle column designed to provide the highest column efficiencies when used with low-dispersive UPLC instrumentation.
  • Offers flexibility to rapidly separate a wide array of compounds.
  • Produces sharper, narrower peaks compared to fully porous particles of similar size.
  • Best column choice for increased resolution, speed, and sensitivity.