Learn More
Waters Corp Waters Column, C18+, 90Å, 1.6 μm, 2.1 mm X 50 mm
Catalog No. 50597083 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C18+ columns are high-efficiency, general-purpose C18 columns with solid-core particle technology. They deliver excellent peak shape for basic analytes at low pH, using sub-2 μm particles for UltraPerformance LC. The C18+ chemistry features Charged Surface Hybrid (CSH) Technology, which imparts a low-level positive charge to the particle surface.
  • Enables new levels of efficiency and superior performance.
  • Improves peak shape, loading capacity, and signal-to-noise for basic compounds in mass spectrometry by using low ionic strength mobile phases.
  • Allows formic acid to substitute ion-pairing reagents while maintaining good peak shape.
  • Facilitates faster separations and greater throughput, enabling more samples or quicker results.
  • Utilizes 1.6 μm solid-core particle technology for UltraPerformance LC capabilities.
  • Lowers analysis and operational costs for laboratories.
Catalog No. 50-597-083 Supplier Waters Corp Supplier No. 186007114
CORTECS C18+ columns are high-efficiency, general-purpose C18 columns with solid-core particle technology. They deliver excellent peak shape for basic analytes at low pH, using sub-2 μm particles for UltraPerformance LC. The C18+ chemistry features Charged Surface Hybrid (CSH) Technology, which imparts a low-level positive charge to the particle surface.
  • Enables new levels of efficiency and superior performance.
  • Improves peak shape, loading capacity, and signal-to-noise for basic compounds in mass spectrometry by using low ionic strength mobile phases.
  • Allows formic acid to substitute ion-pairing reagents while maintaining good peak shape.
  • Facilitates faster separations and greater throughput, enabling more samples or quicker results.
  • Utilizes 1.6 μm solid-core particle technology for UltraPerformance LC capabilities.
  • Lowers analysis and operational costs for laboratories.