Learn More
Waters Corp Waters CORTECS C18 Column, 2.7 μm, 4.6 mm, 150 mm
Supplier: Waters Corp 186007378
The CORTECS C18 Columns are general-purpose, high-efficiency columns based on a solid-core particle that provides balanced retention of acids, bases, and neutrals at low and mid-range pH. This design leads to excellent peak shape for basic analytes at low pH.
- Solid-core particles enhance performance by lowering the van Deemter equation’s A, B, and C terms through more uniform packing, reduced axial diffusion, and improved heat transfer.
- Increased sample throughput with comparable or better peak capacities by increasing the flow rate.
- Shorter column lengths for quicker re-equilibration while maintaining similar peak capacities.
- Reduced lab operation costs by providing speed of analysis without impairing separation performance.
- Designed to deliver high-efficiency results.
- Helps resolve complex mixtures or maintain resolution while increasing throughput.