Learn More
Waters Corp Column, C18+, 90Å, 1.6 µm, 3 mm X 50 mm
Supplier:  Waters Corp 186007119
Encompass_Preferred
CORTECS C18+ columns are general purpose, high efficiency C18 columns based on a solid-core particle that are designed to deliver excellent peak shape for basic analytes at low pH.
  • High efficiency
  • Based on a solid-core particle
  • Designed to deliver excellent peak shape for basic analytes at low pH
Catalog No. 50-239-2677