Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters CORTECS UPLC Column, C18+, 1.6 μm, 2.1 x 30 mm

Supplier:  Waters Corp 176003166

Encompass_Preferred

This CORTECS UPLC Column is a C18+ column with a particle size of 1.6 μm. It features dimensions of 2.1 × 30 mm, offering high performance for various UPLC applications.

  • C18+ column type for versatile separation.
  • Particle size of 1.6 μm for high-resolution separations.
  • Dimensions of 2.1 × 30 mm for efficient analysis.
  • Available in packs of 3.

Catalog No. 50-239-2925


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.