Learn More
Waters Corp CORTECS C18+ Column, 90Å, 2.7 μm, 3 mm x 75 mm
Catalog No. 502391026 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C18+ columns are high-efficiency C18 columns with solid-core particles, optimized for excellent peak shape for basic analytes at low pH. They enhance HPLC/UHPLC system performance and feature a positively charged surface for improved selectivity with acidic, low-ionic strength mobile phases.
  • Solid-core particle technology maximizes efficiency, resolution, and peak capacity.
  • Excellent peak shape for basic analytes at low pH.
  • Positively charged surface modification enhances selectivity for basic compounds with acidic, low-ionic strength mobile phases.
  • High efficiency.
  • Reliable manufacturing ensures consistent performance.
  • Compatible with UPLC and UHPLC systems.
  • Endcapped.
  • Low silanol activity.
  • Spherical particle shape.
Catalog No. 50-239-1026 Supplier Waters Corp Supplier No. 186007401
CORTECS C18+ columns are high-efficiency C18 columns with solid-core particles, optimized for excellent peak shape for basic analytes at low pH. They enhance HPLC/UHPLC system performance and feature a positively charged surface for improved selectivity with acidic, low-ionic strength mobile phases.
  • Solid-core particle technology maximizes efficiency, resolution, and peak capacity.
  • Excellent peak shape for basic analytes at low pH.
  • Positively charged surface modification enhances selectivity for basic compounds with acidic, low-ionic strength mobile phases.
  • High efficiency.
  • Reliable manufacturing ensures consistent performance.
  • Compatible with UPLC and UHPLC systems.
  • Endcapped.
  • Low silanol activity.
  • Spherical particle shape.