Learn More
Waters Corp Column, C18+, 90Å Pore, 2.7 μm, 4.6 mm ID, 50 mm Length
Supplier:  Waters Corp 186007405
Encompass_Preferred
These C18+ columns are general purpose, charged surface, high-efficiency C18 columns with a solid-core particle designed for excellent peak shape for basic analytes at low pH. They enable resolution of complex mixtures and increase throughput.
  • Handle more samples simultaneously while maintaining separation performance.
  • Boost sample throughput by increasing flow rate for higher peak capacities.
  • Withstand pressures up to 18,000 psi (1240 Bar) and temperatures up to 45 °C.
  • Operate in the pH range of 2 to 8.
Catalog No. 50-239-0489