Learn More
Waters Corp CORTECS C18+ Column, 90Å, 2.7 μm, 2.1 x 30 mm, 3/pk
Supplier:  Waters Corp 176003289
Encompass_Preferred
CORTECS C18+ columns are general-purpose, charged-surface, high-efficiency C18 columns based on solid-core particle technology. They deliver excellent peak shape for basic analytes at low pH, efficiently resolving complex mixes and increasing throughput while maintaining separation performance. These columns boost sample throughput by increasing method flow rate, allowing for shorter column lengths and faster separations without sacrificing performance.
  • Excellent peak shape for basic analytes at low pH.
  • Resolves complex mixes, increases throughput.
  • Boosts sample throughput with increased flow rate.
  • Enables shorter column lengths.
  • Provides analysis speed without sacrificing performance.
  • Outstanding selectivity for resolution.
  • Operates 2 to 8 pH range.
  • Max temperature 45 °C.
Catalog No. 50-239-1626