Learn More
Waters Corp CORTECS Column, C18, 2.7 μm, 2.1 x 75 mm
Catalog No. 502390606 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C18 Columns are general purpose, high-efficiency liquid chromatography columns based on a solid-core particle, offering balanced retention for acids, bases, and neutrals at low and mid-range pH. Designed for successful MS adoption and general purpose LC, they maximize efficiency through advanced end-capping processes, providing greater throughput and speed for challenging separations with lasting performance.
  • High-efficiency solid-core particle columns for general purpose LC.
  • Balanced retention of acids, bases, and neutrals over a range of pH.
  • Maximizes performance through advanced end-capping and offers various selectivity options.
  • Provides enhanced throughput and speed for challenging separations.
  • Includes UPLC column protection to extend lifespan and prevent analyte interactions.
Catalog No. 50-239-0606 Supplier Waters Corp Supplier No. 186007366
These C18 Columns are general purpose, high-efficiency liquid chromatography columns based on a solid-core particle, offering balanced retention for acids, bases, and neutrals at low and mid-range pH. Designed for successful MS adoption and general purpose LC, they maximize efficiency through advanced end-capping processes, providing greater throughput and speed for challenging separations with lasting performance.
  • High-efficiency solid-core particle columns for general purpose LC.
  • Balanced retention of acids, bases, and neutrals over a range of pH.
  • Maximizes performance through advanced end-capping and offers various selectivity options.
  • Provides enhanced throughput and speed for challenging separations.
  • Includes UPLC column protection to extend lifespan and prevent analyte interactions.