Learn More
Waters Corp CORTECS C18 Column, 90Å, 2.7 μm, 4.6 mm × 30 mm
Supplier:  Waters Corp 186007374
Encompass_Preferred
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases, and neutrals at low and mid-range pH. The column provides excellent peak shapes for basic analytes at low pH, enables increased sample throughput, and allows for shorter column employment while maintaining peak capacities and shorter re-equilibration times, ultimately lowering lab running expenses.
  • General purpose, high-efficiency column with solid-core particle technology
  • Offers balanced retention for acids, bases, and neutrals at low and mid-range pH
  • Produces excellent peak shapes for basic analytes at low pH
  • Enables increased sample throughput and allows for shorter column use while maintaining peak capacities
  • Solid-core particles provide greater performance by decreasing van Deemter equation terms and enhancing heat transport
Catalog No. 50-239-1081