Learn More
Waters Corp CORTECS C18 Column, 90Å, 2.7 μm, 2.1 mm x 30 mm
Catalog No. 502391612 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • High-efficiency column design
  • Solid-core particle technology
  • Balanced retention for acids, bases, and neutrals at low to mid-range pH
  • Digital access to column documents for less waste
Catalog No. 50-239-1612 Supplier Waters Corp Supplier No. 186007364
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • High-efficiency column design
  • Solid-core particle technology
  • Balanced retention for acids, bases, and neutrals at low to mid-range pH
  • Digital access to column documents for less waste