Learn More
Waters Corp CORTECS UPLC Column, C18, 2.7 μm, 3.0 × 30 mm
Catalog No. 502391688 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS UPLC Column features C18 chemistry, a 2.7 μm particle size, and dimensions of 3.0 × 30 mm. Available as a single column pack, these solid-core particle columns are engineered for high efficiencies with low-dispersive UPLC instrumentation. They produce sharper, narrower peaks than fully porous particles of similar size, making them ideal for enhanced resolution, speed, and sensitivity in separations.
  • Solid-core particle technology
  • Provides high column efficiencies
  • Produces sharper, narrower peaks
  • Designed for increased resolution, speed, and sensitivity
  • C18 chemistry
Catalog No. 50-239-1688 Supplier Waters Corp Supplier No. 186007369
Add to Cart
Add to Cart
The CORTECS UPLC Column features C18 chemistry, a 2.7 μm particle size, and dimensions of 3.0 × 30 mm. Available as a single column pack, these solid-core particle columns are engineered for high efficiencies with low-dispersive UPLC instrumentation. They produce sharper, narrower peaks than fully porous particles of similar size, making them ideal for enhanced resolution, speed, and sensitivity in separations.
  • Solid-core particle technology
  • Provides high column efficiencies
  • Produces sharper, narrower peaks
  • Designed for increased resolution, speed, and sensitivity
  • C18 chemistry