Learn More
Waters Corp UPLC C18 Column, 90 Å, 2.7 μm, 4.6 mm × 50 mm
Catalog No. 502390777 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This CORTECS UPLC C18 Column, supplied in a pack of three, features solid-core particle technology, designed for high-speed applications and producing sharper, narrower peaks. It offers increased resolution, speed, and sensitivity, and is suitable for both HPLC and UHPLC systems. It has a pore size of 90 Å, a particle size of 2.7 μm, and dimensions of 4.6 mm × 50 mm.
  • Features solid-core particle technology for high column efficiencies
  • Produces sharper, narrower peaks compared to fully porous particles of similar size
  • Offers increased resolution, speed, and sensitivity
  • Available in 2.7 μm particles for use with HPLC and UHPLC systems
  • Sold in a pack of three
Catalog No. 50-239-0777 Supplier Waters Corp Supplier No. 176003313
This CORTECS UPLC C18 Column, supplied in a pack of three, features solid-core particle technology, designed for high-speed applications and producing sharper, narrower peaks. It offers increased resolution, speed, and sensitivity, and is suitable for both HPLC and UHPLC systems. It has a pore size of 90 Å, a particle size of 2.7 μm, and dimensions of 4.6 mm × 50 mm.
  • Features solid-core particle technology for high column efficiencies
  • Produces sharper, narrower peaks compared to fully porous particles of similar size
  • Offers increased resolution, speed, and sensitivity
  • Available in 2.7 μm particles for use with HPLC and UHPLC systems
  • Sold in a pack of three