Learn More
Waters Corp Column, C18+, 90Å, 2.7 μm, 4.6 mm, 30 mm
Catalog No. 502390722 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These general purpose, charged surface, high efficiency C18 columns utilize solid-core particles to deliver excellent peak shape for basic analytes at low pH. They enable quicker separations for higher throughput, allowing more concurrent samples and reducing lab operation costs without compromising separation performance.
  • Excellent peak shape for basic analytes at low pH.
  • Quicker separations for higher throughput and reduced lab costs.
  • Solid-core particles improve performance by optimizing the van Deemter equation terms.
  • Improved sample throughput with equivalent or superior peak capacities.
  • Operates across high pressure up to 18,000 psi (1240 Bar).
  • Wide operating pH range from 2 to 8.
  • Compatible with VanGuard cartridges for enhanced protection and extended lifetime.
Catalog No. 50-239-0722 Supplier Waters Corp Supplier No. 186007404
These general purpose, charged surface, high efficiency C18 columns utilize solid-core particles to deliver excellent peak shape for basic analytes at low pH. They enable quicker separations for higher throughput, allowing more concurrent samples and reducing lab operation costs without compromising separation performance.
  • Excellent peak shape for basic analytes at low pH.
  • Quicker separations for higher throughput and reduced lab costs.
  • Solid-core particles improve performance by optimizing the van Deemter equation terms.
  • Improved sample throughput with equivalent or superior peak capacities.
  • Operates across high pressure up to 18,000 psi (1240 Bar).
  • Wide operating pH range from 2 to 8.
  • Compatible with VanGuard cartridges for enhanced protection and extended lifetime.