Learn More
Waters Corp Waters CORTECS C18 Column, 90Å, 2.7 μm, 4.6 mm × 100 mm
Catalog No. 502037970 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C18 Columns are general-purpose, high-efficiency columns based on a solid-core particle, offering balanced retention of acids, bases, and neutrals at low and mid-range pH. This solid-core particle technology provides higher efficiency and increased resolution, enabling increased sample throughput and faster re-equilibration for reduced analysis costs and consistent performance.
  • Achieve high efficiency with solid-core particles for enhanced performance.
  • Ensure balanced retention of acids, bases, and neutrals across low and mid-range pH.
  • Obtain increased resolution for analyzing complex samples.
  • Increase sample throughput by allowing higher flow rates.
  • Reduce analysis costs and lab operational expenses.
  • Benefit from consistent performance due to strict manufacturing vigilance.
  • Solid-core particles improve column efficiency by optimizing van Deemter equation terms.
Catalog No. 50-203-7970 Supplier Waters Corp Supplier No. 186007377
CORTECS C18 Columns are general-purpose, high-efficiency columns based on a solid-core particle, offering balanced retention of acids, bases, and neutrals at low and mid-range pH. This solid-core particle technology provides higher efficiency and increased resolution, enabling increased sample throughput and faster re-equilibration for reduced analysis costs and consistent performance.
  • Achieve high efficiency with solid-core particles for enhanced performance.
  • Ensure balanced retention of acids, bases, and neutrals across low and mid-range pH.
  • Obtain increased resolution for analyzing complex samples.
  • Increase sample throughput by allowing higher flow rates.
  • Reduce analysis costs and lab operational expenses.
  • Benefit from consistent performance due to strict manufacturing vigilance.
  • Solid-core particles improve column efficiency by optimizing van Deemter equation terms.