Learn More
Waters Corp Waters CORTECS C18+ Column, 90Å, 2.7 μm, 3 mm × 100 mm
Supplier:  Waters Corp 186007402
Encompass_Preferred
This general purpose, charged surface, high efficiency C18 column uses a solid-core particle to deliver excellent peak shape for basic analytes at low pH, offering increased sample throughput and better peak capacities.
  • General-purpose, charged surface, high-efficiency solid core C18+ column.
  • Achieves excellent peak shape for basic analytes at low pH.
  • Based on solid-core particles that lower the A, B, and C terms of the van Deemter equation, which improves uniform packing, reduces axial diffusion, and enhances heat transfer.
  • Increases sample throughput with comparable or better peak capacities by raising the flow rate.
  • Allows use of a shorter column length while maintaining similar peak capacities.
Catalog No. NC1538709