Learn More
Waters Corp CORTECS UPLC C18+ Method Validation Kit, 1.6 μm, 90 Å, 2.1 × 75 mm, 3/pk
Catalog No. 502391158 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS UPLC C18+ Method Validation Kit ensures method reproducibility and robustness. It provides three columns from different batches, allowing comprehensive consistency evaluation. This kit utilizes 1.6 μm solid-core particle technology for high efficiency, sharper peaks, and enhanced resolution, speed, and sensitivity.
  • Includes three columns from three batches for method evaluation.
  • Ensures batch-to-batch and column-to-column reproducibility.
  • Utilizes 1.6 μm solid-core particle technology for high efficiency.
  • Provides sharper, narrower peaks for increased resolution, speed, and sensitivity.
Catalog No. 50-239-1158 Supplier Waters Corp Supplier No. 186007178
The CORTECS UPLC C18+ Method Validation Kit ensures method reproducibility and robustness. It provides three columns from different batches, allowing comprehensive consistency evaluation. This kit utilizes 1.6 μm solid-core particle technology for high efficiency, sharper peaks, and enhanced resolution, speed, and sensitivity.
  • Includes three columns from three batches for method evaluation.
  • Ensures batch-to-batch and column-to-column reproducibility.
  • Utilizes 1.6 μm solid-core particle technology for high efficiency.
  • Provides sharper, narrower peaks for increased resolution, speed, and sensitivity.