Learn More
Waters Corp CORTECS Column Kit, C18+, 90Å, 2.7 μm, 3 mm × 150 mm, 3 pk
Catalog No. 502392117 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit provides high-efficiency C18 columns with a charged surface, designed for excellent peak shape of basic analytes at low pH. These columns enable faster separations and higher throughput, offering outstanding retention and stability for resolving difficult mixes or improving throughput.
  • Columns use solid-core particles for excellent peak shape at low pH.
  • Solid-core particles enhance performance by decreasing van Deemter equation terms (A, B, C).
  • Shorter column length maintains peak capacities and delivers faster re-equilibration.
  • Faster analysis allows more samples in parallel, lowering laboratory operating costs.
  • Excellent resolution selectivity enables faster method creation through innovative particle-surface changes.
Catalog No. 50-239-2117 Supplier Waters Corp Supplier No. 186007448
This method validation kit provides high-efficiency C18 columns with a charged surface, designed for excellent peak shape of basic analytes at low pH. These columns enable faster separations and higher throughput, offering outstanding retention and stability for resolving difficult mixes or improving throughput.
  • Columns use solid-core particles for excellent peak shape at low pH.
  • Solid-core particles enhance performance by decreasing van Deemter equation terms (A, B, C).
  • Shorter column length maintains peak capacities and delivers faster re-equilibration.
  • Faster analysis allows more samples in parallel, lowering laboratory operating costs.
  • Excellent resolution selectivity enables faster method creation through innovative particle-surface changes.