Learn More
Waters Corp Method Validation Kit, CORTECS C18+, 90 Å, 2.7 μm
Catalog No. 502392840 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Method Validation Kit features CORTECS C18+ phase with a 90 Å pore size, 2.7 μm particle size, and 3 x 30 mm column dimensions, ideal for robust method development and validation.
  • Method validation kit type
  • CORTECS C18+ phase
  • 90 Å pore size
  • 2.7 μm particle size
  • 3.0 mm ID
  • 30 mm length
  • C18 phase type
  • USP L1 compliant
Catalog No. 50-239-2840 Supplier Waters Corp Supplier No. 186007444
This Method Validation Kit features CORTECS C18+ phase with a 90 Å pore size, 2.7 μm particle size, and 3 x 30 mm column dimensions, ideal for robust method development and validation.
  • Method validation kit type
  • CORTECS C18+ phase
  • 90 Å pore size
  • 2.7 μm particle size
  • 3.0 mm ID
  • 30 mm length
  • C18 phase type
  • USP L1 compliant