Learn More
Waters Corp CORTECS C18+ Column, 2.7 μm, 4.6 mm ID, 30 mm Length, Method Validation Kit
Catalog No. 502392901 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C18+ columns are general purpose, charged surface, high efficiency C18 columns based on a solid-core particle that is designed to deliver excellent peak shape for basic analytes at low pH.
  • Reversed-phase separation mode.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Temperature limits of 45 °C.
  • Maximum pressure of 18000 psi (1240 Bar).
  • Endcapped for enhanced performance.
  • C18+ bonding technology.
  • Low silanol activity.
  • Spherical particle shape.
  • Parker-style endfitting type.
  • Pore diameter of 90 Å.
  • Solid core particle technology.
  • USP Classification L1.
  • Carbon load of 5.7 %.
  • Designed to deliver excellent peak shape for basic analytes at low pH.
Catalog No. 50-239-2901 Supplier Waters Corp Supplier No. 186007449
CORTECS C18+ columns are general purpose, charged surface, high efficiency C18 columns based on a solid-core particle that is designed to deliver excellent peak shape for basic analytes at low pH.
  • Reversed-phase separation mode.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Temperature limits of 45 °C.
  • Maximum pressure of 18000 psi (1240 Bar).
  • Endcapped for enhanced performance.
  • C18+ bonding technology.
  • Low silanol activity.
  • Spherical particle shape.
  • Parker-style endfitting type.
  • Pore diameter of 90 Å.
  • Solid core particle technology.
  • USP Classification L1.
  • Carbon load of 5.7 %.
  • Designed to deliver excellent peak shape for basic analytes at low pH.