Learn More
Waters Corp CORTECS C18+ Method Validation Kit, 1.6 μm, 3 x 30 mm
Catalog No. 502390656 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These general-purpose, high-efficiency C18 columns are based on a solid-core particle and are designed to deliver excellent peak shape for basic analytes at low pH.
  • General purpose, high efficiency C18 columns
  • Based on a solid-core particle
  • Designed to deliver excellent peak shape for basic analytes at low pH
Catalog No. 50-239-0656 Supplier Waters Corp Supplier No. 186007181
These general-purpose, high-efficiency C18 columns are based on a solid-core particle and are designed to deliver excellent peak shape for basic analytes at low pH.
  • General purpose, high efficiency C18 columns
  • Based on a solid-core particle
  • Designed to deliver excellent peak shape for basic analytes at low pH