Learn More
Waters Corp CORTECS C18+ Method Validation Kit, 1.6 μm, 2.1 mm x 50 mm
Catalog No. 502392773 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C18+ columns are general purpose, high efficiency C18 columns based on a solid-core particle that are designed to deliver excellent peak shape for basic analytes at low pH.
  • High efficiency C18 columns
  • Based on a solid-core particle
  • Designed to deliver excellent peak shape for basic analytes at low pH
  • Solid-core particle technology
  • Low silanol activity
  • Spherical particle shape
Catalog No. 50-239-2773 Supplier Waters Corp Supplier No. 186007177
CORTECS C18+ columns are general purpose, high efficiency C18 columns based on a solid-core particle that are designed to deliver excellent peak shape for basic analytes at low pH.
  • High efficiency C18 columns
  • Based on a solid-core particle
  • Designed to deliver excellent peak shape for basic analytes at low pH
  • Solid-core particle technology
  • Low silanol activity
  • Spherical particle shape