Learn More
Waters Corp CORTECS UPLC C18 Method Validation Kit, 3.0 × 30 mm, 1.6 μm, 3 Columns
Catalog No. 502392116 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS UPLC C18 Method Validation Kit (MVK) is designed to assess the reproducibility of a chromatographic column's performance, ensuring the long-term reliability and robustness of analytical methods. This kit contains three C18 columns with a particle size of 1.6 μm and dimensions of 3.0 × 30 mm.
  • Contains three columns, one from each of three different batches, for evaluating method quality, reliability, and consistency.
  • Assesses the reproducibility of chromatographic column performance.
  • Ensures batch-to-batch and column-to-column reproducibility through highly controlled manufacturing processes.
  • Features 3.0 × 30 mm column dimensions.
  • Equipped with 1.6 μm particle size.
Catalog No. 50-239-2116 Supplier Waters Corp Supplier No. 186007161
The CORTECS UPLC C18 Method Validation Kit (MVK) is designed to assess the reproducibility of a chromatographic column's performance, ensuring the long-term reliability and robustness of analytical methods. This kit contains three C18 columns with a particle size of 1.6 μm and dimensions of 3.0 × 30 mm.
  • Contains three columns, one from each of three different batches, for evaluating method quality, reliability, and consistency.
  • Assesses the reproducibility of chromatographic column performance.
  • Ensures batch-to-batch and column-to-column reproducibility through highly controlled manufacturing processes.
  • Features 3.0 × 30 mm column dimensions.
  • Equipped with 1.6 μm particle size.