Learn More
Waters Corp CORTECS Method Validation Kit, C18, 1.6 μm, 3 x 75 mm
Catalog No. 502392577 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS C18 Method Validation Kit provides high-efficiency chromatographic separations, utilizing solid-core particle columns for balanced retention of acids, bases, and neutrals across low to mid-range pH. This kit contains three columns (3 mm × 75 mm L, 1.6 μm spherical silica, 90 Å pore size) compatible with UPLC and UHPLC systems, ensuring reliable performance up to 18000 psi (1240 Bar).
  • High-efficiency columns with solid-core particle technology.
  • Ensures balanced retention for acids, bases, and neutrals in low to mid-range pH.
  • Includes three columns from different batches for method quality and consistency evaluation.
  • Optimized for UPLC instrumentation, producing sharper, narrower peaks for increased resolution and sensitivity.
  • Certificate of Analysis (COAs) available online.
Catalog No. 50-239-2577 Supplier Waters Corp Supplier No. 186007163
The CORTECS C18 Method Validation Kit provides high-efficiency chromatographic separations, utilizing solid-core particle columns for balanced retention of acids, bases, and neutrals across low to mid-range pH. This kit contains three columns (3 mm × 75 mm L, 1.6 μm spherical silica, 90 Å pore size) compatible with UPLC and UHPLC systems, ensuring reliable performance up to 18000 psi (1240 Bar).
  • High-efficiency columns with solid-core particle technology.
  • Ensures balanced retention for acids, bases, and neutrals in low to mid-range pH.
  • Includes three columns from different batches for method quality and consistency evaluation.
  • Optimized for UPLC instrumentation, producing sharper, narrower peaks for increased resolution and sensitivity.
  • Certificate of Analysis (COAs) available online.