Learn More
Waters Corp CORTECS C18 Method Validation Kit, 90Å, 1.6 μm, 3 mm x 50 mm
Catalog No. 502392672 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C18 columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • Column kits
  • C18 chemistry
  • Reversed-phase separation mode
  • Silica particle substrate
  • 2 to 8 pH range
  • 18000 psi maximum pressure (1240 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 1.6 μm particle size
  • 90 Å pore size
  • Method validation kit format
  • 100 surface area
  • UPLC, UHPLC system compatibility
  • Solid core particle technology
  • L1 USP classification
  • 3 mm inner diameter
  • 50 mm length
  • 6.6% carbon load
  • eCord enabled
  • 3 units per package
  • Parker-style endfitting
Catalog No. 50-239-2672 Supplier Waters Corp Supplier No. 186007162
CORTECS C18 columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • Column kits
  • C18 chemistry
  • Reversed-phase separation mode
  • Silica particle substrate
  • 2 to 8 pH range
  • 18000 psi maximum pressure (1240 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 1.6 μm particle size
  • 90 Å pore size
  • Method validation kit format
  • 100 surface area
  • UPLC, UHPLC system compatibility
  • Solid core particle technology
  • L1 USP classification
  • 3 mm inner diameter
  • 50 mm length
  • 6.6% carbon load
  • eCord enabled
  • 3 units per package
  • Parker-style endfitting