Learn More
Waters Corp CORTECS UPLC C18 Method Validation Kit, 1.6 μm, 2.1 × 30 mm, 3/pk
Catalog No. 502392802 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS UPLC C18 Method Validation Kit is designed to assess the reproducibility of chromatographic column performance. It helps evaluate the quality, reliability, and consistency of analytical methods by providing three columns from different batches of material. These columns utilize sub-2-μm, solid-core particle technology for high efficiency and sharper peaks in UPLC applications.
  • Kit contains three columns from three batches.
  • Evaluates analytical method quality, reliability, and consistency.
  • Uses sub-2-μm solid-core particle technology.
  • Provides high column efficiencies for UPLC.
  • Results in sharper, narrower peaks.
  • Ensures batch-to-batch and column-to-column reproducibility.
Catalog No. 50-239-2802 Supplier Waters Corp Supplier No. 186007156
The CORTECS UPLC C18 Method Validation Kit is designed to assess the reproducibility of chromatographic column performance. It helps evaluate the quality, reliability, and consistency of analytical methods by providing three columns from different batches of material. These columns utilize sub-2-μm, solid-core particle technology for high efficiency and sharper peaks in UPLC applications.
  • Kit contains three columns from three batches.
  • Evaluates analytical method quality, reliability, and consistency.
  • Uses sub-2-μm solid-core particle technology.
  • Provides high column efficiencies for UPLC.
  • Results in sharper, narrower peaks.
  • Ensures batch-to-batch and column-to-column reproducibility.