Learn More
Waters Corp Method Validation Kit, C18, 90Å Pore Size, 2.7 μm Particle Size, 3 mm x 30 mm
Catalog No. 502391043 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • High-efficiency columns provide efficient separations.
  • Solid-core particle technology contributes to high efficiency.
  • Balanced retention offers balanced retention for acids, bases, and neutrals.
  • Versatile pH range is effective at low and mid-range pH conditions.
Catalog No. 50-239-1043 Supplier Waters Corp Supplier No. 186007414
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • High-efficiency columns provide efficient separations.
  • Solid-core particle technology contributes to high efficiency.
  • Balanced retention offers balanced retention for acids, bases, and neutrals.
  • Versatile pH range is effective at low and mid-range pH conditions.