Learn More
Waters Corp CORTECS C18 Method Validation Kit, 90Å, 2.7 μm, 2.1 × 100 mm, 3 pk
Catalog No. 502390770 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • Designed for reversed phase separation mode with C18 chemistry.
  • Constructed with silica particle substrate, spherical in shape with low silanol activity.
  • Operates within a pH range of 2 to 8.
  • Withstands maximum pressure of 18000 psi (1240 bar).
  • Compatible with HPLC systems and Parker-style endfitting.
  • Features a pore size of 90Å and a particle size of 2.7 μm.
  • Provided as a method validation kit with an inner diameter of 2.1 mm and a length of 100 mm.
  • Includes 3 units per package.
Catalog No. 50-239-0770 Supplier Waters Corp Supplier No. 186007412
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • Designed for reversed phase separation mode with C18 chemistry.
  • Constructed with silica particle substrate, spherical in shape with low silanol activity.
  • Operates within a pH range of 2 to 8.
  • Withstands maximum pressure of 18000 psi (1240 bar).
  • Compatible with HPLC systems and Parker-style endfitting.
  • Features a pore size of 90Å and a particle size of 2.7 μm.
  • Provided as a method validation kit with an inner diameter of 2.1 mm and a length of 100 mm.
  • Includes 3 units per package.