Learn More
Waters Corp CORTECS VanGuard Cartridge, C18+, 90Å Pore, 2.7 μm Particle
Supplier:  Waters Corp 186007687
Encompass_Preferred
CORTECS C18+ VanGuard Cartridges are designed to prolong the lifespan and maintain the performance of analytical columns. They achieve this by effectively removing particulate contamination from the mobile phase stream. This particular cartridge is specifically optimized to safeguard all 4.6 mm I.D. CORTECS C18+ analytical columns that utilize 2.7 μm sorbent particles.
  • Extends analytical column lifetime
  • Maintains analytical column performance
  • Removes particulate contamination from the mobile phase stream
  • Optimized for 4.6 mm I.D. analytical columns
  • Compatible with 2.7 μm sorbent particles
Catalog No. 50-239-0725