Learn More
Waters Corp CORTECS C18 VanGuard Cartridge, 90 Å, 2.7 μm, 3.9 mm x 5 mm
Catalog No. 502038152 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This VanGuard cartridge extends analytical column lifetime and enhances performance by removing particulate contamination from the mobile phase. Optimized for 4.6 mm I.D. analytical columns with 2.7 μm sorbent particles, it ensures consistent results across configurations. Featuring C18 chemistry, 90 Å pore size, and 3.9 mm inner diameter by 5 mm length, it contributes to longer life, higher throughput, and speed.
  • Extends analytical column lifetime and enhances performance.
  • Optimized for 4.6 mm I.D. analytical columns with 2.7 μm sorbent particles.
  • Ensures consistent results and seamless transfer.
  • Achieves higher throughput and speed.
  • Withstands operating pressures up to 18000 psi (1240 Bar).
  • Provides balanced retention for acids, bases, and neutrals.
Catalog No. 50-203-8152 Supplier Waters Corp Supplier No. 186007684
This VanGuard cartridge extends analytical column lifetime and enhances performance by removing particulate contamination from the mobile phase. Optimized for 4.6 mm I.D. analytical columns with 2.7 μm sorbent particles, it ensures consistent results across configurations. Featuring C18 chemistry, 90 Å pore size, and 3.9 mm inner diameter by 5 mm length, it contributes to longer life, higher throughput, and speed.
  • Extends analytical column lifetime and enhances performance.
  • Optimized for 4.6 mm I.D. analytical columns with 2.7 μm sorbent particles.
  • Ensures consistent results and seamless transfer.
  • Achieves higher throughput and speed.
  • Withstands operating pressures up to 18000 psi (1240 Bar).
  • Provides balanced retention for acids, bases, and neutrals.