Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp CORTECS C8 Column, 90Å, 1.6 μm, 2.1 mm x 75 mm

Catalog No. 502392938 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

CORTECS C8 columns are designed for general purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and, consequently, less retentive compared to C18 phases, making it suitable for certain assays.

  • Chemistry: C8
  • Separation mode: Reversed Phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45°C
  • Maximum pressure: 18000 psi (1240 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Particle shape: Spherical
  • Particle size: 1.6 μm
  • Endfitting type: Parker-style
  • Pore size: 90 Å
  • Format: Column
  • Surface area: 100
  • System compatibility: UPLC, UHPLC
  • Particle technology: Solid Core
  • USP classification: L7
  • Inner diameter: 2.1 mm
  • Length: 75 mm
  • Carbon load: 4.5%
  • eCord: Yes
  • Units per package: 3 pk

Catalog No. 50-239-2938 Supplier Waters Corp Supplier No. 176003831
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CORTECS C8 columns are designed for general purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and, consequently, less retentive compared to C18 phases, making it suitable for certain assays.

  • Chemistry: C8
  • Separation mode: Reversed Phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45°C
  • Maximum pressure: 18000 psi (1240 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Particle shape: Spherical
  • Particle size: 1.6 μm
  • Endfitting type: Parker-style
  • Pore size: 90 Å
  • Format: Column
  • Surface area: 100
  • System compatibility: UPLC, UHPLC
  • Particle technology: Solid Core
  • USP classification: L7
  • Inner diameter: 2.1 mm
  • Length: 75 mm
  • Carbon load: 4.5%
  • eCord: Yes
  • Units per package: 3 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.