Learn More
Waters Corp CORTECS UPLC Column, C8, 1.6 μm, 2.1 x 30 mm
Catalog No. 502390739 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS UPLC 1.6 μm Solid-Core Particle Columns are considered the performance standard. The sub-2-μm, solid-core particle technology offers the highest column efficiencies when used with low-dispersive UPLC instrumentation.
  • Seven distinct chemistries available in either reversed-phase or HILIC, providing flexibility for rapidly separating a broad range of compounds.
  • Produce sharper, narrower peaks compared to fully porous particles of similar size.
  • Optimal choice for increased resolution, speed, and sensitivity.
  • Also available with 2.7 μm particles for use with HPLC and UHPLC systems.
Catalog No. 50-239-0739 Supplier Waters Corp Supplier No. 186008398
CORTECS UPLC 1.6 μm Solid-Core Particle Columns are considered the performance standard. The sub-2-μm, solid-core particle technology offers the highest column efficiencies when used with low-dispersive UPLC instrumentation.
  • Seven distinct chemistries available in either reversed-phase or HILIC, providing flexibility for rapidly separating a broad range of compounds.
  • Produce sharper, narrower peaks compared to fully porous particles of similar size.
  • Optimal choice for increased resolution, speed, and sensitivity.
  • Also available with 2.7 μm particles for use with HPLC and UHPLC systems.