Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, C8, 90Å, 1.6 μm, 3 x 150 mm

Catalog No. 502390990 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These C8 columns are designed for general purpose method development under low pH test conditions. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to C18 phases.

  • Separation mode: Reversed phase
  • Particle size: 1.6 μm
  • Pore size: 90 Å
  • Inner diameter: 3 mm
  • Length: 150 mm
  • pH range: 2 to 8 pH
  • Maximum pressure: 18000 psi (1240 Bar)
  • Temperature limits: 45 °C
  • Particle technology: Solid core
  • Units per package: 3 pk

Catalog No. 50-239-0990 Supplier Waters Corp Supplier No. 176003838
May include imposed supplier surcharges.
Add to Cart
Add to Cart

These C8 columns are designed for general purpose method development under low pH test conditions. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to C18 phases.

  • Separation mode: Reversed phase
  • Particle size: 1.6 μm
  • Pore size: 90 Å
  • Inner diameter: 3 mm
  • Length: 150 mm
  • pH range: 2 to 8 pH
  • Maximum pressure: 18000 psi (1240 Bar)
  • Temperature limits: 45 °C
  • Particle technology: Solid core
  • Units per package: 3 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.