Learn More
Waters Corp Column, C8, 90Å, 1.6 μm, 2.1 mm X 150 mm
Catalog No. 502391810 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C8 columns are designed for general purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and, therefore, less retentive compared to C18 phases, which can be preferred for certain assays.
  • Designed for general purpose method development under low pH test conditions.
  • C8 ligand is less hydrophobic and less retentive compared to C18 phases.
  • Suitable for assays where less retention is preferred.
Catalog No. 50-239-1810 Supplier Waters Corp Supplier No. 176003833
CORTECS C8 columns are designed for general purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and, therefore, less retentive compared to C18 phases, which can be preferred for certain assays.
  • Designed for general purpose method development under low pH test conditions.
  • C8 ligand is less hydrophobic and less retentive compared to C18 phases.
  • Suitable for assays where less retention is preferred.