Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp CORTECS C8 Column, 90Å, 1.6 μm, 3 mm X 50 mm

Catalog No. NC3842859 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

CORTECS C8 columns are utilized for general purpose method development under low pH test conditions. The C8 ligand is favored for certain assays due to its reduced hydrophobicity, resulting in less retention compared to C18 phases. These columns are suitable for specific assays where a less hydrophobic and less retentive phase than C18 is advantageous.

  • Chemistry: C8
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • Particle shape: Spherical
  • Particle technology: Solid core
  • Endcapped: Yes
  • Silanol activity: Low
  • eCord: Yes
  • C8 ligand offers reduced hydrophobicity and retention compared to C18 phases for specific assays

Catalog No. NC3842859 Supplier Waters Corp Supplier No. 186008409
May include imposed supplier surcharges.
Add to Cart
Add to Cart

CORTECS C8 columns are utilized for general purpose method development under low pH test conditions. The C8 ligand is favored for certain assays due to its reduced hydrophobicity, resulting in less retention compared to C18 phases. These columns are suitable for specific assays where a less hydrophobic and less retentive phase than C18 is advantageous.

  • Chemistry: C8
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • Particle shape: Spherical
  • Particle technology: Solid core
  • Endcapped: Yes
  • Silanol activity: Low
  • eCord: Yes
  • C8 ligand offers reduced hydrophobicity and retention compared to C18 phases for specific assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.