Learn More
Waters Corp CORTECS C8 Column, 90 Å, 2.7 μm, 3 mm × 50 mm
Catalog No. 502392764 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This CORTECS C8 column features 2.7 μm solid-core particle technology with a 90 Å pore size, delivering high column efficiencies. It is designed for rapidly separating a wide range of compounds and is compatible with both HPLC and UHPLC systems. This product is available in a 3-pack.
  • Utilizes 2.7 μm solid-core particle technology.
  • Provides high column efficiencies.
  • Offers flexibility for rapidly separating a wide range of compounds.
  • Produces sharper, narrower peaks for enhanced resolution, speed, and sensitivity.
  • Compatible with HPLC and UHPLC systems.
  • Available in a pack of three.
Catalog No. 50-239-2764 Supplier Waters Corp Supplier No. 176003810
This CORTECS C8 column features 2.7 μm solid-core particle technology with a 90 Å pore size, delivering high column efficiencies. It is designed for rapidly separating a wide range of compounds and is compatible with both HPLC and UHPLC systems. This product is available in a 3-pack.
  • Utilizes 2.7 μm solid-core particle technology.
  • Provides high column efficiencies.
  • Offers flexibility for rapidly separating a wide range of compounds.
  • Produces sharper, narrower peaks for enhanced resolution, speed, and sensitivity.
  • Compatible with HPLC and UHPLC systems.
  • Available in a pack of three.