Learn More
Waters Corp CORTECS UPLC Column, C8, 2.7 μm, 3.0 x 100 mm
Catalog No. 502392822 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS UPLC C8 Column utilizes solid-core particle technology to deliver high column efficiencies when used with low-dispersive UPLC instrumentation. These columns are designed to produce sharper, narrower peaks, leading to increased resolution, speed, and sensitivity in separations.
  • Utilizes solid-core particle technology
  • Provides high column efficiencies
  • Produces sharper, narrower peaks
  • Offers increased resolution, speed, and sensitivity
  • Features C8 ligand
  • Endcapped
  • USP Class L7
Catalog No. 50-239-2822 Supplier Waters Corp Supplier No. 176003812
The CORTECS UPLC C8 Column utilizes solid-core particle technology to deliver high column efficiencies when used with low-dispersive UPLC instrumentation. These columns are designed to produce sharper, narrower peaks, leading to increased resolution, speed, and sensitivity in separations.
  • Utilizes solid-core particle technology
  • Provides high column efficiencies
  • Produces sharper, narrower peaks
  • Offers increased resolution, speed, and sensitivity
  • Features C8 ligand
  • Endcapped
  • USP Class L7