Learn More
Waters Corp Column, C8, 90 Å, 2.7 μm, 3 mm x 50 mm
Catalog No. 502390800 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C8 columns are for general purpose method development under low pH conditions. Their C8 ligand is less hydrophobic and retentive than C18 phases, preferred for certain assays. They offer high resolution and efficiency, suitable for separating hydrophobic substances. Can withstand lower back pressures, achieving great throughput and rapid technique development. Solid-core silica technology allows for quick use, cleaning, and reuse.
  • For general purpose method development under low pH conditions.
  • C8 ligand is less hydrophobic and retentive than C18 phases.
  • Offers high resolution and column efficiency.
  • Suitable for separating hydrophobic substances.
  • Withstands lower back pressures.
  • Achieves great throughput and rapid technique development.
  • Solid-core silica technology for quick use, cleaning, and reuse.
Catalog No. 50-239-0800 Supplier Waters Corp Supplier No. 186008359
These C8 columns are for general purpose method development under low pH conditions. Their C8 ligand is less hydrophobic and retentive than C18 phases, preferred for certain assays. They offer high resolution and efficiency, suitable for separating hydrophobic substances. Can withstand lower back pressures, achieving great throughput and rapid technique development. Solid-core silica technology allows for quick use, cleaning, and reuse.
  • For general purpose method development under low pH conditions.
  • C8 ligand is less hydrophobic and retentive than C18 phases.
  • Offers high resolution and column efficiency.
  • Suitable for separating hydrophobic substances.
  • Withstands lower back pressures.
  • Achieves great throughput and rapid technique development.
  • Solid-core silica technology for quick use, cleaning, and reuse.