Learn More
Waters Corp CORTECS C8 Column, 90 Angstrom, 2.7 micrometer, 4.6 x 100 mm
Catalog No. 502390794 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C8 columns are reversed-phase silica columns designed for general purpose method development under low pH conditions, offering high resolution and efficiency for consistent and reliable findings in liquid chromatography.
  • Optimized for low pH method development with C8 ligand, less retentive than C18 phases.
  • Provides high resolution and column efficiency for challenging samples.
  • Ensures lower back pressures, excellent throughput, and quick technique improvement.
  • Features solid-core technology for quick usage, cleaning, and reuse.
  • Ideal for separating extremely hydrophobic substances in UPLC/UHPLC systems.
  • Facilitates various chemical separations due to lower hydrophobicity.
  • Suitable for researchers transitioning to new column technology or diverse applications.
Catalog No. 50-239-0794 Supplier Waters Corp Supplier No. 186008371
CORTECS C8 columns are reversed-phase silica columns designed for general purpose method development under low pH conditions, offering high resolution and efficiency for consistent and reliable findings in liquid chromatography.
  • Optimized for low pH method development with C8 ligand, less retentive than C18 phases.
  • Provides high resolution and column efficiency for challenging samples.
  • Ensures lower back pressures, excellent throughput, and quick technique improvement.
  • Features solid-core technology for quick usage, cleaning, and reuse.
  • Ideal for separating extremely hydrophobic substances in UPLC/UHPLC systems.
  • Facilitates various chemical separations due to lower hydrophobicity.
  • Suitable for researchers transitioning to new column technology or diverse applications.