Learn More
Waters Corp Waters CORTECS C8 Column, 1.6 μm, 90 Å, 2.1 mm x 100 mm
Catalog No. 502224203 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C8 columns are designed for general purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and less retentive than C18 phases, making it ideal for specific assays and separating extremely hydrophobic substances. Featuring solid-core technology, these columns offer higher resolution and efficiency, supporting rapid method development with low back pressures and high throughput.
  • Offers higher resolution and column efficiency.
  • Suitable for developing general liquid chromatography methods.
  • Separates extremely hydrophobic substances due to lower hydrophobicity than C18 phases.
  • Utilizes solid-core technology, allowing for quick use, cleaning, and reuse.
  • Operates with low back pressures, achieving high throughput and rapid method development.
  • Provides a pH limit range of 2 to 8.
  • Withstands a maximum pressure of 18,000 psi (1240 Bar).
Catalog No. 50-222-4203 Supplier Waters Corp Supplier No. 186008401
CORTECS C8 columns are designed for general purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and less retentive than C18 phases, making it ideal for specific assays and separating extremely hydrophobic substances. Featuring solid-core technology, these columns offer higher resolution and efficiency, supporting rapid method development with low back pressures and high throughput.
  • Offers higher resolution and column efficiency.
  • Suitable for developing general liquid chromatography methods.
  • Separates extremely hydrophobic substances due to lower hydrophobicity than C18 phases.
  • Utilizes solid-core technology, allowing for quick use, cleaning, and reuse.
  • Operates with low back pressures, achieving high throughput and rapid method development.
  • Provides a pH limit range of 2 to 8.
  • Withstands a maximum pressure of 18,000 psi (1240 Bar).