Learn More
Waters Corp Waters Column, C8, 90Å, 2.7 μm, 2.1 mm, 50 mm
Catalog No. 502038526 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
C8 columns are designed for general purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and thus less retentive compared to C18 phases, making it preferred for some assays. This column offers greater resolution and column efficiency for liquid chromatography, with each UPLC lab equipment designed for exceptional efficiency across various tests to ensure consistent results.
  • Less hydrophobic than C18 phases, simplifying associated hydrophobic chemical separations.
  • Designed for reduced back pressures while maintaining high throughput and technique development speed.
  • Provides exceptional efficiency across a wide range of tests.
  • Offers greater resolution and column efficiency.
  • Solid-core technology allows for rapid utilization, cleaning, and reuse.
  • Suitable for general purpose method development under low pH test conditions.
Catalog No. 50-203-8526 Supplier Waters Corp Supplier No. 186008349
C8 columns are designed for general purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and thus less retentive compared to C18 phases, making it preferred for some assays. This column offers greater resolution and column efficiency for liquid chromatography, with each UPLC lab equipment designed for exceptional efficiency across various tests to ensure consistent results.
  • Less hydrophobic than C18 phases, simplifying associated hydrophobic chemical separations.
  • Designed for reduced back pressures while maintaining high throughput and technique development speed.
  • Provides exceptional efficiency across a wide range of tests.
  • Offers greater resolution and column efficiency.
  • Solid-core technology allows for rapid utilization, cleaning, and reuse.
  • Suitable for general purpose method development under low pH test conditions.