Learn More
Waters Corp WATERS Column, C8, 90Å, 2.7 μm, 2.1 mm x 100 mm
Catalog No. 502038528 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C8 columns are designed for general purpose method development under low pH test conditions. These columns are less hydrophobic and less retentive compared to C18 phases, making the C8 ligand a preferred choice for certain assays. They offer superior resolution and efficiency across a wide range of tests, ensuring dependable results.
  • Designed for method development in low pH environments.
  • C8 ligand chemistry is less hydrophobic and retentive than C18 phases.
  • Offers superior resolution and efficiency across various sample complexities.
  • Enables low back pressure, quick throughput, and rapid technique development.
  • Ideal for separating very hydrophobic materials.
  • Solid-core silica technology allows for quick utilization and reuse.
  • Supports efficient method development with diverse solvents.
Catalog No. 50-203-8528 Supplier Waters Corp Supplier No. 186008351
CORTECS C8 columns are designed for general purpose method development under low pH test conditions. These columns are less hydrophobic and less retentive compared to C18 phases, making the C8 ligand a preferred choice for certain assays. They offer superior resolution and efficiency across a wide range of tests, ensuring dependable results.
  • Designed for method development in low pH environments.
  • C8 ligand chemistry is less hydrophobic and retentive than C18 phases.
  • Offers superior resolution and efficiency across various sample complexities.
  • Enables low back pressure, quick throughput, and rapid technique development.
  • Ideal for separating very hydrophobic materials.
  • Solid-core silica technology allows for quick utilization and reuse.
  • Supports efficient method development with diverse solvents.