Learn More
Waters Corp Waters CORTECS Column, C8, 90Å, 2.7 μm, 3 mm x 100 mm
Supplier: Waters Corp 186008361
These columns are designed for general purpose method development under low pH test conditions. The C8 ligand is less hydrophobic than C18 phases, making it preferred for some assays, and delivers sharper, narrower peaks for increased resolution, speed, and sensitivity.
- Chemistry: C8; separation mode: reversed phase.
- Particle substrate: Silica; solid core technology.
- Particle size: 2.7 μm; pore size: 90 Å.
- Dimensions: 3 mm inner diameter x 100 mm length.
- pH range: 2 to 8; temperature limit: 45 °C.
- Maximum pressure: 18000 psi (1240 Bar).
- Endcapped with low silanol activity.
- Compatible with UPLC and UHPLC systems.
- Online documentation available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.