Learn More
Waters Corp HPLC Column Cortecs Method Validation Kit, C8, 50 mm, 2.1 mm ID
Catalog No. 502391491 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes a Cortecs column, designed for high-performance liquid chromatography. It features a specific C8 chemistry and precise dimensions for accurate analytical separations.
  • Cortecs column description
  • C8 (RP8, Octyle) package specification
  • 50 mm length
  • 2.1 mm inner diameter
  • 1.6 μm particle size
  • 90 Å pore size
  • Reversed phase mode
  • 2 to 8 pH range
  • Low silanol activity
  • 100 m2/g surface area
  • Endcapped
  • Spherical particles
  • 4.5% carbon load
Catalog No. 50-239-1491 Supplier Waters Corp Supplier No. 186008404
This method validation kit includes a Cortecs column, designed for high-performance liquid chromatography. It features a specific C8 chemistry and precise dimensions for accurate analytical separations.
  • Cortecs column description
  • C8 (RP8, Octyle) package specification
  • 50 mm length
  • 2.1 mm inner diameter
  • 1.6 μm particle size
  • 90 Å pore size
  • Reversed phase mode
  • 2 to 8 pH range
  • Low silanol activity
  • 100 m2/g surface area
  • Endcapped
  • Spherical particles
  • 4.5% carbon load